accessibility menu, dialog, popup

UserName

O

CAS RN 882-59-7

IUPAC Name: 2,2-diphenyloxirane
Synonyms: 2,2-diphenyloxirane
Molecular Weight (g/mol): 196.25
Molecular Formula: C14H12O
InChi Key: PRLJMHVNHLTQJJ-UHFFFAOYSA-N
SMILES: C1OC1(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results