accessibility menu, dialog, popup

UserName

CAS RN 88335-93-7

CH3OOOHO

CAS RN 88335-93-7

IUPAC Name: 6-(methoxycarbonyl)cyclohex-3-ene-1-carboxylic acid
Synonyms: 6-(methoxycarbonyl)cyclohex-3-ene-1-carboxylic acid
Molecular Weight (g/mol): 184.19
Molecular Formula: C9H12O4
InChi Key: MYYLMIDEMAPSGH-UHFFFAOYNA-N
SMILES: COC(=O)C1CC=CCC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results