missing translation for 'onlineSavingsMsg'
Learn More

(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester, 98%

Product Code. 11475720
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475720 5 g 5g
11465720 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475720 Supplier Thermo Scientific Alfa Aesar Supplier No. H25950.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 88335-93-7 | C9H12O4 | 184.19 g/mol

(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester is used in organic synthesis as a starting material and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester is used in organic synthesis as a starting material and intermediate.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents, strong acid and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88335-93-7
Molecular Formula C9H12O4
Molecular Weight (g/mol) 184.19
MDL Number MFCD00757425,MFCD00075490
InChI Key MYYLMIDEMAPSGH-UHFFFAOYNA-N
Synonym 1s,2r-1-methyl cis-1,2,3,6-tetrahydrophthalate, 1r,6s-6-methoxycarbonyl cyclohex-3-enecarboxylic acid, 1r,6s-6-methoxycarbonylcyclohex-3-ene-1-carboxylic acid, 1r,6s-6-methoxycarbonyl cyclohex-3-ene-1-carboxylic acid, 1s,2r-1-methyl cis-4-cyclohexene-1,2-dicarboxylate, 1r,6s-6-methoxycarbonyl-1-cyclohex-3-enecarboxylic acid, 1s,2r-1-methylcis-1,2,3,6-tetrahydrophthalate, 2-hydrogen 1-methyl 1s,2r-1,2,3,6-tetrahydrophthalate, 1 r,6s-6-methoxycarbonyl-3-cyclohexene-1-carboxylic acid, 1r, 2s-2-methoxycarbonylcyclohex-4-ene-1-carboxylic acid
PubChem CID 5325907
IUPAC Name (1R,6S)-6-methoxycarbonylcyclohex-3-ene-1-carboxylic acid
SMILES COC(=O)C1CC=CCC1C(O)=O

Specifications

Melting Point 64°C to 67°C
Quantity 5 g
Beilstein 5262096
Solubility Information Insoluble in water.
Formula Weight 184.19
Percent Purity 98%
Chemical Name or Material (1S,2R)-cis-4-Cyclohexene-1,2-dicarboxylic acid 1-monomethyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.