accessibility menu, dialog, popup

UserName

BNHNOHOH

CAS RN 885068-10-0

IUPAC Name: (1H-indazol-6-yl)boronic acid
Synonyms: 1H-indazol-6-ylboronic acid
Molecular Weight (g/mol): 161.96
Molecular Formula: C7H7BN2O2
InChi Key: ZKNLCHWRWRYPGG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C2C=NNC2=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results