missing translation for 'onlineSavingsMsg'
Learn More

1H-Indazol-6-ylboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 10748668 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10748668 1 g 1g
10768478 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10748668 Supplier Thermo Scientific Acros Supplier No. 431020010
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 885068-10-0
Molecular Formula C7H7BN2O2
Molecular Weight (g/mol) 161.96
MDL Number MFCD04972419
InChI Key ZKNLCHWRWRYPGG-UHFFFAOYSA-N
Synonym 6-indazolyboronic acid, 1h-indazol-6-yl-6-boronic acid, 1h-indazole-6-boronic acid, indazole-6-boronic acid, 1h-indazol-6-yl boronic acid, 6-indazolboronic acid, boronic acid, 1h-indazol-6-yl, 1h-indazole-6-boronicacid, 6-indazoleboronicacid, zlchem 1076
PubChem CID 24728617
IUPAC Name 1H-indazol-6-ylboronic acid
SMILES OB(O)C1=CC=C2C=NNC2=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 180.0°C to 184.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 161.96
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 1H-Indazol-6-ylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 1H-Indazol-6-ylboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.