accessibility menu, dialog, popup

UserName

CAS RN 886498-30-2

H3COFOOHF

CAS RN 886498-30-2

IUPAC Name: 2,6-difluoro-3-methoxybenzoic acid
Synonyms: 2,6-difluoro-3-methoxybenzoic acid
Molecular Weight (g/mol): 188.13
Molecular Formula: C8H6F2O3
InChi Key: YYZBRPSJLASNKL-UHFFFAOYSA-N
SMILES: COC1=C(F)C(C(O)=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results