accessibility menu, dialog, popup

UserName

H3COFNF

CAS RN 886498-35-7

IUPAC Name: 2,6-difluoro-3-methoxybenzonitrile
Synonyms: 2,6-difluoro-3-methoxybenzonitrile
Molecular Weight (g/mol): 169.13
Molecular Formula: C8H5F2NO
InChi Key: BXVCAGRBLOMHBH-UHFFFAOYSA-N
SMILES: COC1=C(F)C(C#N)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results