accessibility menu, dialog, popup

UserName

CAS RN 886498-60-8

H3COFOHF

CAS RN 886498-60-8

IUPAC Name: 2,6-difluoro-3-methoxyphenol
Synonyms: 2,6-difluoro-3-methoxyphenol
Molecular Weight (g/mol): 160.12
Molecular Formula: C7H6F2O2
InChi Key: UOVAWRHEABQHGR-UHFFFAOYSA-N
SMILES: COC1=C(F)C(O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results