accessibility menu, dialog, popup

UserName

H3COFOHF

CAS RN 886498-93-7

IUPAC Name: 2,6-difluoro-4-methoxyphenol
Synonyms: 2,6-difluoro-4-methoxyphenol
Molecular Weight (g/mol): 160.12
Molecular Formula: C7H6F2O2
InChi Key: MGCPGVDPXCHXGO-UHFFFAOYSA-N
SMILES: COC1=CC(F)=C(O)C(F)=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results