accessibility menu, dialog, popup

UserName

CAS RN 886499-40-7

H3COFOOHCl

CAS RN 886499-40-7

IUPAC Name: 2-chloro-6-fluoro-3-methoxybenzoic acid
Synonyms: 2-chloro-6-fluoro-3-methoxybenzoic acid
Molecular Weight (g/mol): 204.58
Molecular Formula: C8H6ClFO3
InChi Key: UXTMPKFTDKMFOF-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C(C(O)=O)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results