accessibility menu, dialog, popup

UserName

CAS RN 886499-54-3

H3COFBrCl

CAS RN 886499-54-3

IUPAC Name: 2-(bromomethyl)-3-chloro-1-fluoro-4-methoxybenzene
Synonyms: 2-(bromomethyl)-3-chloro-1-fluoro-4-methoxybenzene
Molecular Weight (g/mol): 253.5
Molecular Formula: C8H7BrClFO
InChi Key: BIXHHCSLGATAOQ-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C(CBr)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results