accessibility menu, dialog, popup

UserName

CAS RN 886499-83-8

ClHOBrF

CAS RN 886499-83-8

IUPAC Name: 2-bromo-6-chloro-4-fluorophenol
Synonyms: 2-bromo-6-chloro-4-fluorophenol
Molecular Weight (g/mol): 225.44
Molecular Formula: C6H3BrClFO
InChi Key: CRWILTIFGXJONJ-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(F)C=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results