accessibility menu, dialog, popup

UserName

CAS RN 886499-93-0

IUPAC Name: 4-bromo-3-(trifluoromethoxy)phenol
Synonyms: 4-bromo-3-(trifluoromethoxy)phenol
Molecular Weight (g/mol): 257.01
Molecular Formula: C7H4BrF3O2
InChi Key: PGJZBAVEKVWHIO-UHFFFAOYSA-N
SMILES: OC1=CC(OC(F)(F)F)=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results