accessibility menu, dialog, popup

UserName

CAS RN 886501-26-4

OHOFFFF

CAS RN 886501-26-4

IUPAC Name: 4-fluoro-3-(trifluoromethoxy)phenol
Synonyms: 4-fluoro-3-(trifluoromethoxy)phenol
Molecular Weight (g/mol): 196.1
Molecular Formula: C7H4F4O2
InChi Key: FFKOYFSLPUOIJU-UHFFFAOYSA-N
SMILES: OC1=CC(OC(F)(F)F)=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results