accessibility menu, dialog, popup

UserName

CH3OONF

CAS RN 886732-29-2

IUPAC Name: methyl 3-cyano-5-fluorobenzoate
Synonyms: methyl 3-cyano-5-fluorobenzoate
Molecular Weight (g/mol): 179.15
Molecular Formula: C9H6FNO2
InChi Key: YZDWAFGKJVAXAM-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(F)=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results