accessibility menu, dialog, popup

UserName

CAS RN 886763-11-7

IUPAC Name: 2-[2-methyl-4-(trifluoromethoxy)phenyl]acetic acid
Synonyms: [2-methyl-4-(trifluoromethoxy)phenyl]acetic acid
Molecular Weight (g/mol): 234.17
Molecular Formula: C10H9F3O3
InChi Key: QAPHQXYMFGNMCK-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)C=CC(OC(F)(F)F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results