accessibility menu, dialog, popup

UserName

H3CH3CCH3OONNOH

CAS RN 886851-68-9

IUPAC Name: tert-butyl 4-(3-hydroxyphenyl)-1,4-diazepane-1-carboxylate
Synonyms: tert-butyl 4-(3-hydroxyphenyl)-1,4-diazepane-1-carboxylate
Molecular Weight (g/mol): 292.38
Molecular Formula: C16H24N2O3
InChi Key: QDERCJJDCUXJSL-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCCN(CC1)C1=CC(O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results