missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-(3-hydroxyphenyl)perhydro-1,4-diazepine-1-carboxylate, 95%, Thermo Scientific™

Product Code. 11346895
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 11346895

Brand: Thermo Scientific Maybridge MO07901EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 886851-68-9
Molecular Formula C16H24N2O3
Molecular Weight (g/mol) 292.38
MDL Number MFCD08690329
InChI Key QDERCJJDCUXJSL-UHFFFAOYSA-N
Synonym tert-butyl 4-3-hydroxyphenyl perhydro-1,4-diazepine-1-carboxylate, tert-butyl 4-3-hydroxyphenyl-1,4-diazepane-1-carboxylate, 1h-1,4-diazepine-1-carboxylicacid, hexahydro-4-3-hydroxyphenyl-, 1,1-dimethylethyl ester, 1h-1,4-diazepine-1-carboxylicacid,hexahydro-4-3-hydroxyphenyl-,1,1-dimethylethyl ester
PubChem CID 18525947
IUPAC Name tert-butyl 4-(3-hydroxyphenyl)-1,4-diazepane-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCN(CC1)C1=CC(O)=CC=C1

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 292.38
Percent Purity 95%
Chemical Name or Material Tert-butyl 4-(3-hydroxyphenyl)perhydro-1, 4-diazepine-1-carboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.