accessibility menu, dialog, popup

UserName

CAS RN 888070-05-1

CH3OONN

CAS RN 888070-05-1

IUPAC Name: methyl 2-(piperidin-1-yl)pyridine-4-carboxylate
Synonyms: methyl 2-(piperidin-1-yl)pyridine-4-carboxylate
Molecular Weight (g/mol): 220.27
Molecular Formula: C12H16N2O2
InChi Key: CCOBIXWZODVNKL-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(=NC=C1)N1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results