missing translation for 'onlineSavingsMsg'
Learn More

Methyl 2-piperidinoisonicotinate, 97%, Thermo Scientific™

Product Code. 10173124
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
Product Code. Quantity Packaging unitSize
10173124 250 mg Amber Glass Bottle 250mg
10142794 1 g Amber Glass Bottle 1g
2 options
This item is not returnable. View return policy

Product Code. 10173124

Brand: Thermo Scientific Maybridge CC50322CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 888070-05-1
Molecular Formula C12H16N2O2
Molecular Weight (g/mol) 220.27
MDL Number MFCD08741429
InChI Key CCOBIXWZODVNKL-UHFFFAOYSA-N
Synonym methyl 2-piperidinoisonicotinate, methyl 2-piperidin-1-ylpyridin-4-ylcarboxylate, methyl 2-piperidin-1-yl pyridine-4-carboxylate, methyl 2-piperidin-1-yl isonicotinate, methyl 2-piperidin-1-ylisonicotinate, methyl 2-piperidylpyridine-4-carboxylate, 4-pyridinecarboxylicacid, 2-1-piperidinyl-, methyl ester, 4-pyridinecarboxylicacid,2-1-piperidinyl-,methyl ester
PubChem CID 18525843
IUPAC Name methyl 2-piperidin-1-ylpyridine-4-carboxylate
SMILES COC(=O)C1=CC(=NC=C1)N1CCCCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 220.27
Percent Purity 97%
Chemical Name or Material Methyl 2-piperidinoisonicotinate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.