accessibility menu, dialog, popup

UserName

NH2ClHOO

CAS RN 89-54-3

IUPAC Name: 5-amino-2-chlorobenzoic acid
Synonyms: 5-amino-2-chlorobenzoic acid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: GVCFFVPEOLCYNN-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C(=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)