accessibility menu, dialog, popup

UserName

CAS RN 89-64-5

OHClNOO

CAS RN 89-64-5

IUPAC Name: 4-chloro-2-nitrophenol
Synonyms: 4-chloro-2-nitrophenol
Molecular Weight (g/mol): 173.55
Molecular Formula: C6H4ClNO3
InChi Key: NWSIFTLPLKCTSX-UHFFFAOYSA-N
SMILES: OC1=CC=C(Cl)C=C1[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results