accessibility menu, dialog, popup

UserName

CAS RN 89615-42-9

IUPAC Name: 3,4-bis(benzyloxy)-5-[(benzyloxy)methyl]oxolan-2-ol
Synonyms: 3,4-bis(benzyloxy)-5-[(benzyloxy)methyl]oxolan-2-ol
Molecular Weight (g/mol): 420.51
Molecular Formula: C26H28O5
InChi Key: NAQUAXSCBJPECG-UHFFFAOYNA-N
SMILES: OC1OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results