missing translation for 'onlineSavingsMsg'
Learn More

2,3,5-Tri-O-benzyl-L-arabinofuranose, 98%, Thermo Scientific Chemicals

Product Code. 15467659
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15467659 1 g 1g
15487659 250 mg 250mg
15477659 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15467659 Supplier Thermo Scientific Alfa Aesar Supplier No. J66343.03
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89615-42-9 | Molecular Formula: C26H28O5 | Molecular Weight: 420.505 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89615-42-9
Molecular Formula C26H28O5
Molecular Weight (g/mol) 420.505
MDL Number MFCD00053542
InChI Key NAQUAXSCBJPECG-SSUZURRFSA-N
Synonym 2,3,5-tri-o-benzyl-beta-l-arabinofuranose, 2s,3r,4s,5s-3,4-bis benzyloxy-5-benzyloxy methyl tetrahydrofuran-2-ol, 2,3,5-tri-o-benzyl-l-arabinofuranose, 2s,3r,4s,5s-3,4-bis benzyloxy-5-benzyloxy methyl oxolan-2-ol, 2,3,5-tri-o-benzyl-beta-l-arabino-furanose, b-l-arabinofuranose,2,3,5-tris-o-phenylmethyl
PubChem CID 12933788
IUPAC Name (2S,3R,4S,5S)-3,4-bis(phenylmethoxy)-5-(phenylmethoxymethyl)oxolan-2-ol
SMILES C1=CC=C(C=C1)COCC2C(C(C(O2)O)OCC3=CC=CC=C3)OCC4=CC=CC=C4

Specifications

Color White
Quantity 1 g
Formula Weight 420.5
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,3,5-Tri-O-benzyl-L-arabinofuranose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.