accessibility menu, dialog, popup

UserName

CH3OH3COHO

CAS RN 89639-83-8

IUPAC Name: 4,5-dimethylfuran-2-carboxylic acid
Synonyms: 4,5-dimethylfuran-2-carboxylic acid
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: BNVXYXRSJIWWDX-UHFFFAOYSA-N
SMILES: CC1=C(C)C=C(O1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results