missing translation for 'onlineSavingsMsg'
Learn More

4,5-Dimethyl-2-furoic acid, 97%, Thermo Scientific™

Product Code. 10326834
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10326834 5 g Amber Glass Bottle 5g
10304612 250 mg Amber Glass Bottle 250mg
10314612 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10326834 Supplier Thermo Scientific Maybridge Supplier No. BTB08890DE
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 89639-83-8
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00085031
InChI Key BNVXYXRSJIWWDX-UHFFFAOYSA-N
PubChem CID 2774924
IUPAC Name 4,5-dimethylfuran-2-carboxylic acid
SMILES CC1=C(OC(=C1)C(=O)O)C

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 4,5-dimethyl-2-furoic acid, 2-furancarboxylic acid,4,5-dimethyl, 2-furancarboxylic acid, 4,5-dimethyl, 3-methylbenzofurane, acmc-20lon1, 4,5-dimethylfurancarboxylic acid, 4,5-dimethyl-2-furancarboxylic acid, 4,5-dimethyl-furan-2-carboxylic acid
Formula Weight 140.14
Percent Purity 97%
Chemical Name or Material 4, 5-Dimethyl-2-furoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.