accessibility menu, dialog, popup

UserName

H3CCH3BOHHO

CAS RN 89787-12-2

IUPAC Name: [2-(propan-2-yl)phenyl]boronic acid
Synonyms: 2-isopropylphenylboronic acid
Molecular Weight (g/mol): 164.01
Molecular Formula: C9H13BO2
InChi Key: KTZUVUWIBZMHMC-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=CC=C1B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results