accessibility menu, dialog, popup

UserName

NNCl

CAS RN 898289-48-0

IUPAC Name: 2-[4-(chloromethyl)phenyl]pyrimidine
Synonyms: 2-[4-(chloromethyl)phenyl]pyrimidine
Molecular Weight (g/mol): 204.66
Molecular Formula: C11H9ClN2
InChi Key: ZYZLFVLHAHBXGB-UHFFFAOYSA-N
SMILES: ClCC1=CC=C(C=C1)C1=NC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results