accessibility menu, dialog, popup

UserName

CH3NOOFFF

CAS RN 89976-12-5

IUPAC Name: 1-methyl-4-nitro-2-(trifluoromethyl)benzene
Synonyms: 1-methyl-4-nitro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 205.14
Molecular Formula: C8H6F3NO2
InChi Key: SVQCVQCIZWSPPX-UHFFFAOYSA-N
SMILES: CC1=C(C=C(C=C1)[N+]([O-])=O)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results