accessibility menu, dialog, popup

UserName

NH2

CAS RN 90-41-5

IUPAC Name: [1,1'-biphenyl]-2-amine
Synonyms: 2-aminobiphenyl
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: TWBPWBPGNQWFSJ-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)