missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobiphenyl, 98%

Product Code. 10290983 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10290983 25 g Glass bottle 25g
10765741 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10290983 Supplier Thermo Scientific Acros Supplier No. 103220250
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90-41-5 | C12H11N | 169.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-41-5
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00007701
InChI Key TWBPWBPGNQWFSJ-UHFFFAOYSA-N
Synonym 2-aminobiphenyl, 1,1'-biphenyl-2-amine, 2-biphenylamine, 2-aminodiphenyl, o-aminobiphenyl, o-biphenylamine, biphenyl-2-ylamine, o-aminodiphenyl, o-phenylaniline, biphenyl-2-amine
PubChem CID 7015
IUPAC Name 2-phenylaniline
SMILES NC1=CC=CC=C1C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 48°C to 52°C
Color Purple to Brown
Boiling Point 299°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H4NH2
Quantity 25 g
Beilstein 12, 1317
Formula Weight 169.23
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2-Aminobiphenyl, 98%
Show More Show Less
compliance-icons compliance-icons
Nombre del producto
  • 2-Aminobiphenyl
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
Indicaciones de peligro
  • H351-Suspected of causing cancer.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Consejos de prudencia
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.