CAS RN 9003-05-8
IUPAC Name:
prop-2-enamide
Synonyms:
acrylamide bio-gel P 2 polyacrylamide solution acrylic amide vinyl amide biogel P-100 polyacrylamide resin akrylamid acrylagel K-pam
Molecular Weight (g/mol):
71.08
Molecular Formula:
C3H5NO
InChi Key:
HRPVXLWXLXDGHG-UHFFFAOYSA-N
SMILES:
NC(=O)C=C
1
–
3
of
3
results
1
Poly(acrylamide), granular, non-ionic, approx. M.W. 5 to 6.000.000
CAS: 9003-05-8 Molecular Formula: (C3H5NO)n MDL Number: MFCD00084392 InChI Key: HRPVXLWXLXDGHG-UHFFFAOYSA-N SMILES: NC(=O)C(-*)C-*
2
Polyacrylamide (10% in H2O, Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Polyacrylamide (10% in H2O, Technical Grade), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.