missing translation for 'onlineSavingsMsg'
Learn More

Polyacrylamide (10% in H2O, Technical Grade), TRC

Product Code. 30417520
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30417520 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30417520 Supplier Toronto Research Chemicals Supplier No. TRCP6881501G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Polyacrylamide
CAS 9003-05-8
Molecular Formula (C3H5NO)x
Quantity 1 g
Synonym Acrylamide','polymers (8CI), 2-Propenamide hydrochloride homopolymer, 2300S, 2J, 3330s, 38F, 920MPM, AK, AK (flocculant), AMF, AP 273, ASP 6, Accotrol S 622, Acrylamide homopolymer, Acryperse M 2000A, Alcoflood 1175, American Cyanamid KPAM, American Cyanamid P 250, Aminogen PA, Amol, Aquamid, Aquasorb 3005KB, Aron F 40, BanDrift, Boze Floc N 46BT, CM 303, CM 311, CM 311 (polymer), CPF 802, CS 141, CS 141 (polymer), Calgon 470, Calgon 800, Cogum 20P, Cogum 25H, Colsize WLV, Companion PCT 12, Cosmogel, Cosmotone Z, Criterion TGX Any kD Stain-Free Precast Gel, Cyanamer A 15L, Cytame 5, DALT, Diaclear MA 3000H, Diaclear MN 3000, Diaclear MN 3000H, Discol 4600, Fennopol K 6450, Floc 1403C, Flopam AN 945, Flygtol GB, Formacryl, Formula 358, GPA-u, Gelamide 250, Get-Down, H 106, H 106 (acrylic resin), Hamacoat P 3800, Haricoat, Harmide K 123, Harmide RB 32, Himoloc ND 270, Himoloc OK 507, Himoloc SS 200, Hiset P 700, Hiset P 700SN, Hiset PN 550, Hiset PN 750, Hydraid 6696EZ, IPM 320, Instar NS, Kuricoat C 402, Lamflood 746, MS 4000HSF, Magnafloc E 30, Magnifloc 900N, Marwit SU 4, Marwit SU 8, Meipam M 200, Migunon NS, Mirbane 301, Multifloc 231, Multigel 10/20, Myresin R 10L, Nalco, Nalsize 7541, Nordfloc AN 920SH, Orfloc ON 2, Pamid, Pararokku 2A111, Parasol MG, Percol 2300, Percol 333, Percol 3335, Percol 540, Plex 4847D, Polias 320, Polyacron KR 143, Polyacrylamide, Polyacrylon PD 365R, Polyhall, Polymerset, Polymin, Polysic, Polystron, Polytex A, Pomosist 117, Praestol, Proquim 17-14, Quick Floc, Rheolate 402, Rira Gum SJ 108, S 602, SFA 120, ST 50000, Sanfloc AL 310P, Sanpoly A 520, Santac, Seabetter AD, Sedifloc, Sedipur TF, Sedipur TF 514, Sepaflux CE 5174, Shallol AM 253P, Solidokoll K, Solvitose 433, Stargum AD-S, Stockosorb KL, Stokopol D 2624, Sumifloc FN 10H, Sumirez, Superfloc N, Sursolan P 5, TRP 1044, TY 007, Texapret AM, Tulsepar PNS 1, VN 728, Versicol, Vistowall 300, Viterra II, WIN-NPH, X-Coat P 130C, Zonen A, c-PAGEL
Molecular Weight (g/mol) 101.15
Concentration or Composition (by Analyte or Components) 0.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.