accessibility menu, dialog, popup

UserName

CAS RN 9005-12-3

IUPAC Name: methyl(phenyl)silanone
Synonyms: phenyl dimethicone
Molecular Weight (g/mol): 136.23
Molecular Formula: C7H8OSi
InChi Key: IXKNJYSAWNJQQY-UHFFFAOYSA-N
SMILES: C[Si](=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results