missing translation for 'onlineSavingsMsg'
Learn More

Polyphenylmethylsiloxane, MW 2500-2700

Product Code. 11485340
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485340 1 kg 1kg
11465340 50 g 50g
11475340 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11485340 Supplier Thermo Scientific Alfa Aesar Supplier No. 044569.A1
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9005-12-3 | C7H8OSi | 136.23 g/mol

A biochemical for proteomics research. A novel coating for solid-phase microextraction. A polyphenylmethylsiloxane for use in skincare, suncare, color cosmetic, hair care, and antiperspirant / deodorant applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-12-3
MDL Number MFCD00146731

Specifications

Physical Form Liquid
Molecular Formula (C24H32O16)n
Synonym triacetylcellulose, cellulose triacetate, beta-cellotriose undecaacetate, 2r,3r,4s,5r,6s-4,5-bis acetyloxy-6-2r,3r,4s,5r,6s-4,5,6-tris acetyloxy-2-acetyloxy methyl oxan-3-yl oxy-3-2s,3r,4s,5r,6r-3,4,5-tris acetyloxy-6-acetyloxy methyl oxan-2-yl oxy oxan-2-yl methyl acetate
Solubility Information Insoluble in water.
Quantity 1 kg
InChI Key NNLVGZFZQQXQNW-ADJNRHBOSA-N
SMILES CC(=O)OCC1C(C(C(C(O1)OC2C(OC(C(C2OC(=O)C)OC(=O)C)OC(=O)C)COC(=O)C)OC(=O)C)OC(=O)C)OC3C(C(C(C(O3)COC(=O)C)OC(=O)C)OC(=O)C)OC(=O)C
IUPAC Name [(2R,3R,4S,5R,6S)-4,5-diacetyloxy-3-[(2S,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxy-6-[(2R,3R,4S,5R,6S)-4,5,6-triacetyloxy-2-(acetyloxymethyl)oxan-3-yl]oxyoxan-2-yl]methyl acetate
PubChem CID 44263853
Flash Point 300°C (572°F)
Chemical Name or Material Polyphenylmethylsiloxane, M.W. 2,500 to 2,700
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.