CAS RN 90296-27-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals
CAS: 90296-27-8 Molecular Formula: C8H9ClO2 Molecular Weight (g/mol): 172.61 MDL Number: MFCD02683547 InChI Key: AXXPZAITCCIOIA-UHFFFAOYSA-N Synonym: 4-chloro-2-methoxybenzyl alcohol,4-chloro-2-methoxyphenyl methanol,4-chloro-2-methoxy-phenyl-methanol,benzenemethanol, 4-chloro-2-methoxy,4-chloro-2-methoxybenzyl alcohol 97 PubChem CID: 3549393 IUPAC Name: (4-chloro-2-methoxyphenyl)methanol SMILES: COC1=C(CO)C=CC(Cl)=C1