accessibility menu, dialog, popup

UserName

CAS RN 9036-19-5

CH3CH3H3CCH3H3COOOH

CAS RN 9036-19-5

IUPAC Name: 2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethan-1-ol
Synonyms: 2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethanol
Molecular Weight (g/mol): 294.44
Molecular Formula: C18H30O3
InChi Key: LBCZOTMMGHGTPH-UHFFFAOYSA-N
SMILES: CC(C)(C)CC(C)(C)C1=CC=C(OCCOCCO)C=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results