accessibility menu, dialog, popup

UserName

CAS RN 90397-54-9

IUPAC Name: lanthanum(3+) triethanolate
Synonyms: lanthanum(3+) tris(ethoxide)
Molecular Weight (g/mol): 274.09
Molecular Formula: C6H15LaO3
InChi Key: NPAJGHOZGYPSTK-UHFFFAOYSA-N
SMILES: [La+3].CC[O-].CC[O-].CC[O-]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results