missing translation for 'onlineSavingsMsg'
Learn More

Lanthanum(III) ethoxide, 95% (REO), Thermo Scientific™

Product Code. 11398677
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398677 2 g 2g
11308687 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398677 Supplier Thermo Scientific Alfa Aesar Supplier No. 035791.04

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90397-54-9
Molecular Formula C6H15LaO3
Molecular Weight (g/mol) 274.088
MDL Number MFCD00075070
InChI Key NPAJGHOZGYPSTK-UHFFFAOYSA-N
Synonym lanthanum iii ethoxide, lanthanum triethoxide, lanthanum 3+ tris ethoxide, lanthanum 3+ ion tris ethoxide
PubChem CID 14274587
IUPAC Name ethanolate;lanthanum(3+)
SMILES CC[O-].CC[O-].CC[O-].[La+3]

Specifications

Color White
Linear Formula La(OCH2CH3)3
Quantity 2 g
UN Number UN2925
Sensitivity Moisture sensitive
Formula Weight 274.08
Percent Purity 95%
Physical Form Crystalline
Assay (REO)
Chemical Name or Material Lanthanum(III) ethoxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.