accessibility menu, dialog, popup

UserName

CAS RN 905-62-4

ONN

CAS RN 905-62-4

IUPAC Name: bis(naphthalen-1-yl)-1,3,4-oxadiazole
Synonyms: bis(naphthalen-1-yl)-1,3,4-oxadiazole
Molecular Weight (g/mol): 322.37
Molecular Formula: C22H14N2O
InChi Key: MUNFOTHAFHGRIM-UHFFFAOYSA-N
SMILES: O1C(=NN=C1C1=C2C=CC=CC2=CC=C1)C1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results