missing translation for 'onlineSavingsMsg'
Learn More

2,5-Bis(1-naphthyl)-1,3,4-oxadiazole, 99%, Thermo Scientific™

Product Code. 11377186
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377186 5 g 5g
11367186 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11377186 Supplier Thermo Scientific Alfa Aesar Supplier No. L07215.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter

Denne vare kan ikke returneres. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 905-62-4
Molecular Formula C22H14N2O
Molecular Weight (g/mol) 322.37
MDL Number MFCD00045582
InChI Key MUNFOTHAFHGRIM-UHFFFAOYSA-N
Synonym 2,5-di 1-naphthyl-1,3,4-oxadiazole, 2,5-bis 1-naphthyl-1,3,4-oxadiazole, 2,5-di naphthalen-1-yl-1,3,4-oxadiazole, bis naphthalen-1-yl-1,3,4-oxadiazole, 2,5-di-naphthalen-1-yl-1,3,4 oxadiazole, acmc-209r6s, 2,5-dinaphthyl-1,3,4-oxadiazole, 2,5-di 1-naphtyl-1,3,4-oxadiazole, 2,5-bis 1-naphtyl-1,3,4-oxadiazole, 2,5-bis-4-naphthyl-1,3,4-oxadiazole
PubChem CID 70182
IUPAC Name 2,5-dinaphthalen-1-yl-1,3,4-oxadiazole
SMILES O1C(=NN=C1C1=C2C=CC=CC2=CC=C1)C1=C2C=CC=CC2=CC=C1

Tekniske data

Melting Point 175°C to 178°C
Quantity 5 g
Beilstein 285250
Formula Weight 322.37
Percent Purity 99%
Chemical Name or Material 2,5-Bis(1-naphthyl)-1,3,4-oxadiazole
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.