accessibility menu, dialog, popup

UserName

CAS RN 906352-71-4

CH3NHOO

CAS RN 906352-71-4

IUPAC Name: methyl({[2-(oxan-4-yloxy)phenyl]methyl})amine
Synonyms: methyl({[2-(oxan-4-yloxy)phenyl]methyl})amine
Molecular Weight (g/mol): 221.3
Molecular Formula: C13H19NO2
InChi Key: ZTMRIJRRCHAUNA-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC=C1OC1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results