accessibility menu, dialog, popup

UserName

CAS RN 906353-02-4

CH3NNOHO

CAS RN 906353-02-4

IUPAC Name: {4-[(6-methylpyrazin-2-yl)oxy]phenyl}methanol
Synonyms: {4-[(6-methylpyrazin-2-yl)oxy]phenyl}methanol
Molecular Weight (g/mol): 216.24
Molecular Formula: C12H12N2O2
InChi Key: RNFIYSOVNHRFJW-UHFFFAOYSA-N
SMILES: CC1=CN=CC(OC2=CC=C(CO)C=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results