CAS RN 90717-16-1
CAS RN 90717-16-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-[(2-Chlorophenyl)(methylimino)methyl]cyclopentanol Hydrochloride, TRC
CAS: 90717-16-1 Molecular Formula: C13 H16 Cl N O . Cl H Molecular Weight (g/mol): 274.19 Synonym: Cyclopentanol, 1-[(2-chlorophenyl)(methylimino)methyl]-, hydrochloride (1:1),Cyclopentanol, 1-[(2-chlorophenyl)(methylimino)methyl]-, hydrochloride (9CI),2-Chlorophenyl-1-hydroxy-1-cyclopentyl N-methyl ketimine hydrochloride IUPAC Name: 1-[(E)-C-(2-chlorophenyl)-N-methylcarbonimidoyl]cyclopentan-1-ol;hydrochloride SMILES: Cl.C\N=C(/c1ccccc1Cl)\C2(O)CCCC2