CAS RN 908143-04-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Linezolid Dimer, TRC
CAS: 908143-04-4 Molecular Formula: C30H35F2N5O7 Molecular Weight (g/mol): 615.62 Synonym: N,N-bis[[(5S)-3-[3-fluoro-4-(4-morpholinyl)phenyl]-2-oxo-5-oxazolidinyl]methyl]acetamide,Bis(linezolid),Bis-Linezolid,Linezolid Impurity D IUPAC Name: N,N-bis[[(5S)-3-(3-fluoro-4-morpholin-4-ylphenyl)-2-oxo-1,3-oxazolidin-5-yl]methyl]acetamide SMILES: CC(=O)N(C[C@H]1CN(C(=O)O1)C2=CC(=C(C=C2)N3CCOCC3)F)C[C@H]4CN(C(=O)O4)C5=CC(=C(C=C5)N6CCOCC6)F