accessibility menu, dialog, popup

UserName

MgCl

CAS RN 90878-19-6

IUPAC Name: magnesium(2+) 2-phenylethanide chloride
Synonyms: magnesium(2+) 2-phenylethanide chloride
Molecular Weight (g/mol): 164.92
Molecular Formula: C8H9ClMg
InChi Key: OUTILEHLSPAZIN-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].[CH2-]CC1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results