missing translation for 'onlineSavingsMsg'
Learn More

Phenethylmagnesium chloride, 1M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10143204 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10143204 800 mL AcroSeal™ Glass Bottle 800mL
10041034 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143204 Supplier Thermo Scientific Acros Supplier No. 427808000
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90878-19-6 | Molecular Formula: C8H9ClMg | Molecular Weight: 164.92 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90878-19-6 , 109-99-9
Molecular Formula C8H9ClMg
Molecular Weight (g/mol) 164.92
MDL Number MFCD00192387
InChI Key AZORNQXQALOFHH-UHFFFAOYSA-M
Synonym phenethyl magnesium chloride, phenylethyl magnesium chloride, chloro 2-phenylethyl magnesium, phenethylmagnesium chloride, phenethylmagnesium chloride solution, phenethymagnesium chloride, phenyethylmagnesium chloride, phenylethylmagnesium chloride, 2-phenylethylmagnesium chloride, 2-phenylethyl magnesium chloride
PubChem CID 4671868
IUPAC Name magnesium;ethylbenzene;chloride
SMILES Cl[Mg]CCC1=CC=CC=C1

Specifications

Color Yellow to Brown
Density 0.9540g/mL
Flash Point −17°C
Packaging AcroSeal™ Glass Bottle
Linear Formula C6H5(CH2)2MgCl
Quantity 800 mL
Specific Gravity 0.954
Formula Weight 164.92
Physical Form Liquid
Chemical Name or Material Phenethylmagnesium chloride, 1M solution in THF
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phenethylmagnesium chloride, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.