CAS RN 90965-06-3
IUPAC Name:
1-(diazyn-1-ium-1-yl)-1-(dimethoxyphosphoryl)prop-1-en-2-olate
Synonyms:
seyferth-gilbert homologation
Molecular Weight (g/mol):
192.11
Molecular Formula:
C5H9N2O4P
InChi Key:
SQHSJJGGWYIFCD-UHFFFAOYSA-N
SMILES:
COP(=O)(OC)C([N+]#N)=C(C)[O-]
1
–
4
of
4
results
1
Dimethyl (1-diazo-2-oxopropyl)phosphonate, 95%
CAS: 90965-06-3 Molecular Formula: C5H9N2O4P Molecular Weight (g/mol): 192.11 InChI Key: SQHSJJGGWYIFCD-UHFFFAOYSA-N IUPAC Name: 1-(diazyn-1-ium-1-yl)-1-(dimethoxyphosphoryl)prop-1-en-2-olate SMILES: COP(=O)(OC)C([N+]#N)=C(C)[O-]
2
Dimethyl (1-Diazo-2-oxopropyl)phosphonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dimethyl (1-Diazo-2-oxopropyl)phosphonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Dimethyl (1-Diazo-2-oxopropyl)phosphonate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.