missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl (1-Diazo-2-oxopropyl)phosphonate, TRC

Product Code. 30371411
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30371411 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30371411 Supplier Toronto Research Chemicals Supplier No. TRCD471815250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethyl (1-Diazo-2-oxopropyl)phosphonate
CAS 90965-06-3
Molecular Formula C5H9N2O4P
Quantity 250 mg
Synonym Bestmann-Ohira Reagent, 1-Diazo-1-(dimethyl phosphono)acetone, Bestmann Reagent, Dimethyl (1-Azo-2-oxopropyl)phosphonate, Dimethyl (1-azoacetonyl)phosphonate, Dimethyl (1-Diazo-2-oxopropyl)phosphonate, Dimethyl (acetyldiazomethyl)phosphonate, Ohira-Bestmann Phosphonate Ohira-Bestmann Reagent, Ohira's Reagent, α-Azido-α-(Dimethyl phosphono)acetone
InChI Formula InChI=1S/C5H9N2O4P/c1-4(8)5(7-6)12(9,10-2)11-3/h1-3H3
SMILES O=C(C)C(P(OC)(OC)=O)=[N+]=[N-]
Molecular Weight (g/mol) 192.11
Formula Weight 192.03
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.