CAS RN 910037-02-4
IUPAC Name:
2,3,4,5,6-pentafluorophenyl 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylate
Synonyms:
2,3,4,5,6-pentafluorophenyl 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylate
Molecular Weight (g/mol):
366.26
Molecular Formula:
C14H7F5O4S
InChi Key:
FHURIHOENRMCSL-UHFFFAOYSA-N
SMILES:
FC1=C(F)C(F)=C(OC(=O)C2=C(C=CS2)C2OCCO2)C(F)=C1F
1
–
1
of
1
results
1
Pentafluorophenyl 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylate, 97%, Thermo Scientific™
CAS: 910037-02-4 Molecular Formula: C14H7F5O4S Molecular Weight (g/mol): 366.26 MDL Number: MFCD09702392 InChI Key: FHURIHOENRMCSL-UHFFFAOYSA-N Synonym: pentafluorophenyl 3-1,3-dioxolan-2-yl thiophene-2-carboxylate,2,3,4,5,6-pentafluorophenyl 3-1,3-dioxolan-2-yl thiophene-2-carboxylate,perfluorophenyl 3-1,3-dioxolan-2-yl thiophene-2-carboxylate,2,3,4,5,6-pentakis fluoranyl phenyl 3-1,3-dioxolan-2-yl thiophene-2-carboxylate,3-1,3-dioxolan-2-yl-2-thiophenecarboxylic acid 2,3,4,5,6-pentafluorophenyl ester PubChem CID: 24229594 IUPAC Name: (2,3,4,5,6-pentafluorophenyl) 3-(1,3-dioxolan-2-yl)thiophene-2-carboxylate SMILES: FC1=C(F)C(F)=C(OC(=O)C2=C(C=CS2)C2OCCO2)C(F)=C1F